Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hettich
®
ROTOFIX 32A/Universal 320/320R centrifuge accessories
Products
Manufacturer
Package Size
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z657891
Hettich
®
ROTOFIX 32A/Universal 320/320R centrifuge accessories,
for UNIVERSAL 320; ROTOFIX 32 A Centrifuges, Fixed Angle Rotor, 6 place
Expand
View Pricing
Z631140
Hettich
®
ROTOFIX 32A/Universal 320/320R centrifuge accessories,
Adapter for 15 mL Falcon tubes 17x120 mm, for UNIVERSAL 320
Expand
View Pricing
Z722839
Hettich
®
ROTOFIX 32A/Universal 320/320R centrifuge accessories,
Rotor, 2 place for microplates, for UNIVERSAL 320
Expand
View Pricing
Z722790
Hettich
®
ROTOFIX 32A/Universal 320/320R centrifuge accessories,
Rotor, 4 place, for ROTOFIX 32A and UNIVERSAL 320
Expand
View Pricing
Z722847
Hettich
®
ROTOFIX 32A/Universal 320/320R centrifuge accessories,
Rotor, 6 place for CYTO, swing out, for ROTOFIX 32A and UNIVERSAL 320
Expand
View Pricing
Z722804
Hettich
®
ROTOFIX 32A/Universal 320/320R centrifuge accessories,
Rotor, 8 place without tube holders, for ROTOFIX 32A and UNIVERSAL 320
Expand
View Pricing
Z653381
Hettich
®
ROTOFIX 32A/Universal 320/320R centrifuge accessories,
Adapter for 4-7 mL tubes 13x100 mm, for ROTOFIX 32 and UNIVERSAL 320
Expand
View Pricing