
Hederagenin

Synonym(s):
3,23-Dihydroxyolean-12-en-28-acid, Astrantiagenin E, Caulosapogenin, Hederagenic acid, Hederagenol
Empirical Formula (Hill Notation):
C30H48O4
CAS Number:
Molecular Weight:
472.70
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing