Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
HayeSep
®
Porous Polymer Adsorbent
Products
Brand
Supelco (4)
Manufacturer
Physical Form
Quality Segment
Package Size
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
10301-U
HayeSep
®
Porous Polymer Adsorbent,
matrix HayeSep Q, 80-100 mesh, bottle of 75 cc
Expand
View Pricing
10291
HayeSep
®
Porous Polymer Adsorbent,
matrix HayeSep D, 60-80 mesh, bottle of 75 cc
Expand
View Pricing
10300-U
HayeSep
®
Porous Polymer Adsorbent,
matrix HayeSep Q, 60-80 mesh, bottle of 75 cc
Expand
View Pricing
10295
HayeSep
®
Porous Polymer Adsorbent,
matrix HayeSep N, 80-100 mesh, bottle of 75 cc
Expand
View Pricing
10294
HayeSep
®
Porous Polymer Adsorbent,
matrix HayeSep N, 60-80 mesh, bottle of 75 cc
Expand
View Pricing
10303
HayeSep
®
Porous Polymer Adsorbent,
matrix HayeSep R, 60-80 mesh, bottle of 75 cc
Expand
View Pricing
10283
HayeSep
®
Porous Polymer Adsorbent,
matrix HayeSep A, 80-100 mesh, bottle of 75 cc
Expand
View Pricing
10290
HayeSep
®
Porous Polymer Adsorbent,
matrix HayeSep C, 100-120 mesh, bottle of 75 cc
Expand
View Pricing
10284
HayeSep
®
Porous Polymer Adsorbent,
matrix HayeSep A, 100-120 mesh, bottle of 75 cc
Expand
View Pricing
10286
HayeSep
®
Porous Polymer Adsorbent,
matrix HayeSep B, 80-100 mesh, bottle of 75 cc
Expand
View Pricing
10297
HayeSep
®
Porous Polymer Adsorbent,
matrix HayeSep P, 60-80 mesh, bottle of 75 cc
Expand
View Pricing