Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn
Products
Manufacturer
Physical Form
Diameter (mm)
ID (mm)
Length (m)
Length (mm)
OD (mm)
Filter Products
Showing 1-20 of 96
Page 1 of 5
Page 1 of 5
Compare
Product Number
Description
Pricing
GF79877783
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
10m wire, 1.0mm diameter, hard
Expand
View Pricing
GF99660617
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
foil, 100 x 100mm, 0.15mm thickness, annealed
Expand
View Pricing
GF41221336
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
rod, 200mm x 25mm diameter
Expand
View Pricing
GF50276921
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
foil, 0.2m coil, 0.025mm thickness, as rolled
Expand
View Pricing
GF79919803
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
5m wire, 0.5mm diameter, hard
Expand
View Pricing
GF15482134
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
foil, 50 x 50mm, 0.025mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF15537719
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
rod, 100mm x 12.7mm diameter
Expand
View Pricing
GF63997284
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
foil, 50 x 50mm, 0.15mm thickness, annealed
Expand
View Pricing
GF41952090
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
rod, 100mm x 16mm diameter
Expand
View Pricing
GF52888146
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
200mm tube, 6.35mm outside diameter, 0.9mm wall thickness, 4.55mm inside diameter
Expand
View Pricing
GF68680626
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
200m wire, 0.1mm diameter, hard
Expand
View Pricing
GF62111977
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
foil, 25 x 25mm, 0.15mm thickness, annealed
Expand
View Pricing
GF24451309
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
10m wire, 0.1mm diameter, hard
Expand
View Pricing
GF14287967
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
200mm tube, 3.2mm outside diameter, 0.9mm wall thickness, 1.4mm inside diameter
Expand
View Pricing
GF84723037
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
20m wire, 1.0mm diameter, hard
Expand
View Pricing
GF97131012
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
rod, 50mm x 30mm diameter
Expand
View Pricing
GF66297752
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
rod, 50mm x 12.7mm diameter
Expand
View Pricing
GF19715632
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
100mm tube, 6.35mm outside diameter, 0.9mm wall thickness, 4.55mm inside diameter
Expand
View Pricing
GF70188071
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
1m wire, 1.0mm diameter, hard
Expand
View Pricing
GF58389542
Hastelloy
®
C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn,
500mm tube, 9.5mm outside diameter, 0.9mm wall thickness, 7.7mm inside diameter
Expand
View Pricing
Page 1 of 5
Page 1 of 5