Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hamilton
®
needles for RN syringes
Products
Product Category
needles (8)
Manufacturer
Length (in)
Length (mm)
Volume (µL)
Needle Shape
Package Size
Gauge
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
19136-U
Hamilton
®
needles for RN syringes,
needle size 22s ga, needle L 51 mm (2 in.), bevel tip stainless steel needle, pkg of 6 ea
Expand
View Pricing
19131-U
Hamilton
®
needles for RN syringes,
volume 2.5-100 μL, bevel tip stainless steel needle (point style 2), needle size 26s ga
Expand
View Pricing
19137-U
Hamilton
®
needles for RN syringes,
needle size 22 ga, bevel tip (point style 2), needle L 51 mm (2 in.), for use with 250μL-10mL
Expand
View Pricing
19133-U
Hamilton
®
needles for RN syringes,
needle size 22s ga, bevel tip stainless steel needle, needle L 51 mm (2 in.), pkg of 6 ea
Expand
View Pricing
Z118125
Hamilton
®
needles for RN syringes,
volume 2.5-100 μL, needle size 22 ga, bevel tip, needle L 101.5 mm (4 in.)
Expand
View Pricing
Z288993
Hamilton
®
needles for RN syringes,
volume 2.5-100 μL, needle size 22 ga, bevel tip, needle L 51 mm (2 in.)
Expand
View Pricing
Z118117
Hamilton
®
needles for RN syringes,
needle size 22 ga, bevel tip, needle L 76 mm (3 in.), volume 2.5-100 μL
Expand
View Pricing
Z118109
Hamilton
®
needles for RN syringes,
volume 250-10000 μL, needle size 22 ga, bevel tip, needle L 101.5 mm (4 in.)
Expand
View Pricing