
Guanosine 5′-diphospho-D-mannose sodium salt from Saccharomyces cerevisiae

Synonym(s):
GDP-Man, GDP-mannose
Linear Formula:
C16H23N5O16P2Na2
CAS Number:
Molecular Weight:
649.30
Products
Brand
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing