Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Greiner CELLSTAR
®
96 well plates
Products
Product Category
plates (15)
Brand
Sigma-Aldrich (2)
Manufacturer
Binding Type
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
M0812
Greiner CELLSTAR
®
96 well plates,
flat bottom clear wells
Expand
View Pricing
M0562
Greiner CELLSTAR
®
96 well plates,
flat bottom black polystyrene wells (with micro-clear bottom)
Expand
View Pricing
M0187
Greiner CELLSTAR
®
96 well plates,
flat bottom white wells
Expand
View Pricing
M3687
Greiner CELLSTAR
®
96 well plates,
size 96 wells, flat bottom clear polystyrene wells, lid, case of 60 ea
Expand
View Pricing
M0437
Greiner CELLSTAR
®
96 well plates,
flat bottom white polystyrene wells (with micro-clear bottom)
Expand
View Pricing
M3562
Greiner CELLSTAR
®
96 well plates,
size 96 wells, round bottom clear wells, lid, case of 60
Expand
View Pricing
M9686
Greiner CELLSTAR
®
96 well plates,
V-bottom clear wells
Expand
View Pricing
M0312
Greiner CELLSTAR
®
96 well plates,
flat bottom black polystyrene wells
Expand
View Pricing
M9561
Greiner CELLSTAR
®
96 well plates,
V-bottom clear wells
Expand
View Pricing
M9936
Greiner CELLSTAR
®
96 well plates,
flat bottom black polystyrene wells
Expand
View Pricing
M9436
Greiner CELLSTAR
®
96 well plates,
round bottom clear wells
Expand
View Pricing
M9311
Greiner CELLSTAR
®
96 well plates,
round bottom clear wells
Expand
View Pricing
M0687
Greiner CELLSTAR
®
96 well plates,
flat bottom clear wells
Expand
View Pricing
M1062
Greiner CELLSTAR
®
96 well plates,
flat bottom white polystyrene wells (with micro-clear bottom)
Expand
View Pricing
M0937
Greiner CELLSTAR
®
96 well plates,
flat bottom clear polystyrene wells (with micro-clear bottom)
Expand
View Pricing