Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Greiner 96 well plates, polystyrene
Products
Product Category
plates (7)
Brand
Sigma-Aldrich (3)
Color
Manufacturer
Binding Type
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
M2936
Greiner 96 well plates, polystyrene,
non-sterile
Expand
View Pricing
M3061
Greiner 96 well plates, polystyrene,
flat bottom clear polystyrene wells, sterile
Expand
View Pricing
M2186
Greiner 96 well plates, polystyrene,
round bottom clear wells, non-sterile
Expand
View Pricing
M2311
Greiner 96 well plates, polystyrene,
round bottom clear wells, sterile
Expand
View Pricing
M2686
Greiner 96 well plates, polystyrene,
conical bottom clear wells, non-sterile
Expand
View Pricing
M3186
Greiner 96 well plates, polystyrene,
flat bottom clear polystyrene wells, sterile
Expand
View Pricing
M2811
Greiner 96 well plates, polystyrene,
conical bottom clear wells, sterile
Expand
View Pricing