
Graphene dispersion

Synonym(s):
conductive ink
Linear Formula:
SiCxHyOz
Products
Brand
Sustainability
Boiling Point (°C)
Feature
Physical Form
Chemical Composition
Quality Segment
Particle Size (µm)
Showing 1-9 of 9
Compare
Product Number
Description
Pricing