Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Graduated Valumetric
™
bottles
Products
Product Category
bottles (16)
Brand
Sigma-Aldrich (2)
Capacity (mL)
Diameter (mm)
Height (mm)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
Z547557
Graduated Valumetric
™
bottles,
polypropylene cap (with poly-vinyl liner), capacity 125 mL (4 oz)
Expand
View Pricing
Z547530
Graduated Valumetric
™
bottles,
polypropylene cap (with poly-vinyl liner), capacity 30 mL (1 oz)
Expand
View Pricing
Z551031
Graduated Valumetric
™
bottles,
clear soda-lime glass bottle, capacity 250 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 24 ea
Expand
View Pricing
Z551198
Graduated Valumetric
™
bottles,
black phenolic cap (with Poly-Seal
®
liner), capacity 60 mL (2 oz)
Expand
View Pricing
Z551023
Graduated Valumetric
™
bottles,
clear soda-lime glass bottle, capacity 125 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 24 ea
Expand
View Pricing
Z550981
Graduated Valumetric
™
bottles,
polypropylene cap (with PTFE-faced liner), capacity 30 mL (1 oz)
Expand
View Pricing
Z550965
Graduated Valumetric
™
bottles,
polypropylene cap (with poly-vinyl liner), capacity 250 mL (8 oz)
Expand
View Pricing
Z551007
Graduated Valumetric
™
bottles,
clear soda-lime glass bottle, capacity 60 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 48 ea
Expand
View Pricing
Z551228
Graduated Valumetric
™
bottles,
black phenolic cap (with rubber liner), capacity 60 mL (2 oz)
Expand
View Pricing
Z551058
Graduated Valumetric
™
bottles,
clear soda-lime glass bottle, capacity 500 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 24 ea
Expand
View Pricing
Z551201
Graduated Valumetric
™
bottles,
black phenolic cap (with rubber liner), capacity 30 mL (1 oz)
Expand
View Pricing
Z551171
Graduated Valumetric
™
bottles,
black phenolic cap (with Poly-Seal
®
liner), capacity 30 mL (1 oz)
Expand
View Pricing
Z550973
Graduated Valumetric
™
bottles,
polypropylene cap (with poly-vinyl liner), capacity 500 mL (16 oz)
Expand
View Pricing
Z551236
Graduated Valumetric
™
bottles,
black phenolic cap (with rubber liner), capacity 125 mL (4 oz)
Expand
View Pricing
Z551244
Graduated Valumetric
™
bottles,
black phenolic cap (with rubber liner), capacity 250 mL (8 oz)
Expand
View Pricing
Z547549
Graduated Valumetric
™
bottles,
polypropylene cap (with poly-vinyl liner), capacity 60 mL (2 oz)
Expand
View Pricing