Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs SecureSeal
™
hybridization chambers
Products
Manufacturer
Diameter (mm)
Length (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-20 of 25
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GBL621505
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 8, well diam. × depth 9 mm × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL623504
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 1, well L × W × depth 40 mm × 22 mm × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL621502
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 1, well diam. × depth 13 mm × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL621101
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 8, well diam. × depth 9 mm × 1 mm, port diam. 1.5 mm
Expand
View Pricing
GBL621202
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 8, well diam. × depth 9 mm × 2.6 mm, port diam. 1.5 mm
Expand
View Pricing
GBL621501
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 1, well diam. × depth 20 mm × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL622510
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 16, well L × W × depth 7 mm × 7 mm × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL622514
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 6, well L × W × depth 20 mm × 9.8 mm × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL623503
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 1, well L × W × depth from septum to bottom of cone 22 mm × 22 mm × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL622506
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 1, well L × W × depth 53 mm × 22 mm × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL621503
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 8, well L × L × depth 7 mm × 7 mm × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL621211
Grace Bio-Labs SecureSeal
™
hybridization chambers,
well diam. × depth 20 mm × 1.8 mm, wells, 1, port diam. 1.5 mm
Expand
View Pricing
GBL621506
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 1, well L × W × depth 60 mm × 22 mm × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL621201
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 8, well diam. × depth 9 mm × 1.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL621301
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 1, well diam. × depth 20 mm × 2.6 mm, port diam. 1.5 mm
Expand
View Pricing
GBL621111
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 1, well diam. × depth 20 mm × 1 mm, port diam. 1.5 mm
Expand
View Pricing
GBL622508
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 3, well L × W × depth 21 mm × 20 mm × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL622512
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 2, well L × W × depth 34.5 mm × 21.5 mm × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL623507
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 1, well L × W × depth 60 mm × 22 mL/ft × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
GBL622513
Grace Bio-Labs SecureSeal
™
hybridization chambers,
wells, 1, well L × W × depth 60 mm × 21 mm × 0.8 mm, port diam. 1.5 mm
Expand
View Pricing
Page 1 of 2
Page 1 of 2