Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs SecureSeal
™
adhesive sheets
Products
Product Category
adhesives (5)
Manufacturer
Length (in)
Color
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GBL620001
Grace Bio-Labs SecureSeal
™
adhesive sheets,
thickness 0.12 mm , double sided, size 7.25 in. × 10 in.
Expand
View Pricing
GBL620002
Grace Bio-Labs SecureSeal
™
adhesive sheets,
thickness 0.24 mm , double sided, diam. 7.25 in. × 10 in.
Expand
View Pricing
GBL620003
Grace Bio-Labs SecureSeal
™
adhesive sheets,
thickness 0.36 mm , double sided, size 7.25 in. × 10 in.
Expand
View Pricing
GBL620005
Grace Bio-Labs SecureSeal
™
adhesive sheets,
thickness 0.60 mm, Double Sided, L × W 7 in. × 10 in.
Expand
View Pricing
GBL620004
Grace Bio-Labs SecureSeal
™
adhesive sheets,
thickness 0.48 mm , double sided, size 7.25 in. × 10 in.
Expand
View Pricing