Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side
Products
Manufacturer
Length (mm)
Surface Area cm²
Color
Package Size
Sterility
Filter Products
Showing 1-20 of 31
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GBL665504
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 1 (Round, 20mm diameter, 0.6mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665104
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 1 (Round, 20mm diameter, 0.9mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665507
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 1 (Round, 13mm diameter, 0.6mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665217
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 16 (Square, 7mm x 7mm x 1.7mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665506
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 12 (Round, 4.5mm diameter, 0.6mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665501
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 8 (Round, 9mm diameter, 0.6mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665108
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 24 (Round, 2mm diameter, 0.9mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665301
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
(Adhesive on one side)
Expand
View Pricing
GBL665206
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 12 (ound, 4.5mm diameter, 1.7mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665503
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 1 (Rounded rectangle, 19mm x 32mm x 0.6mm depth), (Adhesive one side)
Expand
View Pricing
GBL665103
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 1 (Rounded rectangle, 19mm x 32mm x 0.9mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665201
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 8 (Round, 9mm diameter, 1.7mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665106
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 12 (Round, 4.5mm diameter, 0.9mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665207
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 1 (Round, 13mm diameter, 1.7mm depth), (Adhesive one side)
Expand
View Pricing
GBL665303
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 1 (Rounded rectangle, 19mm x 32mm x 2.5mm depth), (Adhesive one side)
Expand
View Pricing
GBL665304
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 1 (Round, 20mm diameter, 2.5mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665216
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 10 (Square, 7mm x 7mm x 1.7mm depth), (Adhesive one side)
Expand
View Pricing
GBL665101
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 8 (Round, 9mm diameter, 0.9mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665215
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 8 (Square, 7mm x 7mm x 1.7mm depth), (Adhesive on one side)
Expand
View Pricing
GBL665208
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side,
wells, 24 (Round, 2mm diameter, 1.7mm depth), (Adhesive on one side)
Expand
View Pricing
Page 1 of 2
Page 1 of 2