Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs FlexWell
™
removable incubation chambers
Products
Manufacturer
Length (mm)
Surface Area cm²
Color
Package Size
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
GBL204064
Grace Bio-Labs FlexWell
™
removable incubation chambers,
L × W × thickness 3.5 mm × 3.5 mm × 1.8 mm, black silicone, adhesive one side, wells 64
Expand
View Pricing
GBL204916
Grace Bio-Labs FlexWell
™
removable incubation chambers,
L × W × thickness 6.5 mm × 6.5 mm × 3.2 mm, clear silicone, adhesive one side, wells 16
Expand
View Pricing
GBL204908
Grace Bio-Labs FlexWell
™
removable incubation chambers,
L × W × thickness 6.5 mm × 6.5 mm × 3.2 mm, clear silicone, adhesive one side, wells, 8
Expand
View Pricing