Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs CultureWell
™
chambered coverglass
Products
Product Category
slides (1)
Manufacturer
Diameter (mm)
Length (mm)
Surface Area cm²
Volume (µL)
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GBL103350
Grace Bio-Labs CultureWell
™
chambered coverglass,
wells, 50, well diam. × thickness 3 mm × 1 mm, well volume 3-10 μL
Expand
View Pricing
GBL103380
Grace Bio-Labs CultureWell
™
chambered coverglass,
wells, 8, well diam. × thickness 6 mm × 1 mm, well volume 15-30 μL
Expand
View Pricing
GBL103330
Grace Bio-Labs CultureWell
™
chambered coverglass,
wells, 3, well diam. × thickness 9.5 mm × 1 mm, well volume 300-500 μL
Expand
View Pricing
GBL103340
Grace Bio-Labs CultureWell
™
chambered coverglass,
wells, 4, diam. × thickness 9 mm × 1 mm, volume 50-100 μL
Expand
View Pricing
GBL103310
Grace Bio-Labs CultureWell
™
chambered coverglass,
wells, 2, diam. × thickness 15 mm × 1 mm, volume 250-400 μL
Expand
View Pricing
GBL103320
Grace Bio-Labs CultureWell
™
chambered coverglass,
wells, 2, well diam. × thickness 15 mm × 1.5 mm, well volume 300-500 μL
Expand
View Pricing