Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gold coated silica nanoparticles
Linear Formula:
SiO
2
@AuNP
CAS Number:
7631-86-9
Products
Brand
Sigma-Aldrich (4)
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
926949
Gold coated silica nanoparticles,
1 wt. % in water, 200 nm (non-porous silica), 20 nm (gold nanoparticles)
Expand
View Pricing
926957
Gold coated silica nanoparticles,
1 wt. % in water, 400 nm (non-porous silica), 20 nm (gold nanoparticles)
Expand
View Pricing
926973
Gold coated silica nanoparticles,
1 wt. % in water, 800 nm (non porous silica), 20 nm (gold nanoparticles)
Expand
View Pricing
926965
Gold coated silica nanoparticles,
1 wt. % in water, 600 nm (non-porous silica), 20 nm (gold nanoparticles)
Expand
View Pricing