![Glycine tert-butyl ester hydrochloride 97%](/deepweb/assets/sigmaaldrich/product/structures/224/515/44ad9b0f-811b-424c-8caa-13360403ec9a/640/44ad9b0f-811b-424c-8caa-13360403ec9a.png)
Glycine tert-butyl ester hydrochloride
![Glycine tert-butyl ester hydrochloride 97%](/deepweb/assets/sigmaaldrich/product/structures/224/515/44ad9b0f-811b-424c-8caa-13360403ec9a/640/44ad9b0f-811b-424c-8caa-13360403ec9a.png)
Synonym(s):
tert-Butyl aminoacetate hydrochloride
Linear Formula:
H2NCH2CO2C(CH3)3 · HCl
CAS Number:
Molecular Weight:
167.63
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing