Skip to Content
Merck
Glycine, Molecular Biology Grade Absorbance (260 nm and 280 nm): ≤0.01.

Glycine, Molecular Biology Grade

Glycine, Molecular Biology Grade Absorbance (260 nm and 280 nm): ≤0.01.
Synonym(s):
Glycine, Aminoacetic acid, Aminoethanoic acid, Glycocoll
Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07

Products

Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
pK Value
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing