Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glyceryl tripalmitate
Synonym(s):
Glycerol tripalmitate, Tripalmitin
Linear Formula:
[CH
3
(CH
2
)
14
COOCH
2
]
2
CHOCO(CH
2
)
14
CH
3
CAS Number:
555-44-2
Molecular Weight:
807.32
EC Number:
209-098-1
Products
Product Category
glycerides (2)
lipids (2)
metabolites (1)
Brand
Sigma-Aldrich (2)
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
T5888
Glyceryl tripalmitate,
≥99%
Expand
View Pricing
T8127
Glyceryl tripalmitate,
≥85%
Expand
View Pricing