Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glutamic-Pyruvic Transaminase from porcine heart
Synonym(s):
L-Alanine: 2-Oxoglutarate Aminotransferase, Alanine Aminotransferase (catalytic concentration), GPT
CAS Number:
9000-86-6
EC Number:
232-561-4
Products
Brand
Sigma-Aldrich (2)
Biological Source
Color
Physical Form
Tissue
Chemical Composition
Quality Segment
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
G8255
Glutamic-Pyruvic Transaminase from porcine heart,
lyophilized powder, ≥75 units/mg protein
Expand
View Pricing
G9880
Glutamic-Pyruvic Transaminase from porcine heart,
ammonium sulfate suspension, ≥75 units/mg protein
Expand
View Pricing