Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glucose-6-phosphate Dehydrogenase from
Leuconostoc mesenteroides
Synonym(s):
G-6-P-DH
CAS Number:
9001-40-5
EC Number:
232-602-6
Products
Brand
Sigma-Aldrich (4)
Biological Source
Physical Form
Recombinant Host
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
G8404
Glucose-6-phosphate Dehydrogenase from
Leuconostoc mesenteroides
,
recombinant, expressed in
E. coli
, ammonium sulfate suspension, ≥550 units/mg protein (biuret)
Expand
View Pricing
G5885
Glucose-6-phosphate Dehydrogenase from
Leuconostoc mesenteroides
,
lyophilized powder, >= 550 units/mg protein (biuret)
Expand
View Pricing
G5760
Glucose-6-phosphate Dehydrogenase from
Leuconostoc mesenteroides
,
Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret
Expand
View Pricing
G8529
Glucose-6-phosphate Dehydrogenase from
Leuconostoc mesenteroides
,
recombinant, expressed in
E. coli
, lyophilized powder, ≥550 units/mg protein (biuret)
Expand
View Pricing