Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glass Wool
EC Number:
266-046-0
Products
Brand
Supelco (6)
Agency
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
20411
Glass Wool,
Silanized, pkg of 50 g
Expand
View Pricing
20410
Glass Wool,
Silanized, pkg of 250 g
Expand
View Pricing
20384
Glass Wool,
Non-Treated, pkg of 50 g
Expand
View Pricing
20409
Glass Wool,
Pesticide Grade (Silanized), pkg of 10 g
Expand
View Pricing
20383
Glass Wool,
Phosphoric Acid Treated, pkg of 50 g
Expand
View Pricing
21688-U
Glass Wool,
Pesticide Grade (Silanized), pkg of 100 g
Expand
View Pricing