Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gentian Violet
Synonym(s):
Crystal Violet, Basic Violet 3, Gentian Violet, Hexamethylpararosaniline chloride, Methyl Violet 10B
Linear Formula:
C
25
H
30
N
3
Cl
CAS Number:
548-62-9
Molecular Weight:
407.98
EC Number:
208-953-6
Products
Brand
Sigma-Aldrich (1)
Formula Weight
Manufacturer
Melting Point (°C)
Activity Spectrum
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Mode of Action
API Family
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1290002
Gentian Violet,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
G2039
Gentian Violet,
meets USP testing specifications
Expand
View Pricing