Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gel cutting tips
Products
Product Category
pipette tips (9)
Manufacturer
OD (mm)
Color
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
AXYTGL1165
Gel cutting tips,
size 1.1 mm × 6.5 mm, pkg of 10x250tips/cs
Expand
View Pricing
AXYTGL200RD57R
Gel cutting tips,
round, racked, O.D. 0.57 mm, size 200 μL, pkg of 2x200tips/cs
Expand
View Pricing
AXYTGL200RD57RS
Gel cutting tips,
O.D. 0.57 mm, round, racked, size 200 μL, pkg of 2x200tips/cs, sterile
Expand
View Pricing
AXYTGL1165R
Gel cutting tips,
size 1.1 mm × 6.5 mm, pkg of 5x480tips/cs, racked
Expand
View Pricing
AXYTGL200FT37R
Gel cutting tips,
flat, racked, O.D. 0.37 mm, size 200 μL, pkg of 2x200tips/cs
Expand
View Pricing
AXYTGL10FT17R
Gel cutting tips,
flat, racked, O.D. 0.17 mm, size 10 μL, pkg of 2x200tips/cs
Expand
View Pricing
AXYTGL1140
Gel cutting tips,
size 1.1 mm × 4 mm, pkg of 10x250tips/cs
Expand
View Pricing
AXYTGL1140R
Gel cutting tips,
size 1.1 mm × 4 mm, pkg of 5x480tips/cs, racked
Expand
View Pricing
AXYTGL200FT37RS
Gel cutting tips,
O.D. 0.37 mm, flat, racked, size 200 μL, pkg of 2x200tips/cs, sterile
Expand
View Pricing