Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gas Sampling Bulb
Products
Brand
Supelco (7)
Quality Segment
Application
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
22161
Gas Sampling Bulb,
size 125 mL, PTFE stopcock
Expand
View Pricing
22162-U
Gas Sampling Bulb,
size 250 mL, PTFE stopcock
Expand
View Pricing
22144-U
Gas Sampling Bulb,
size 1000 mL, glass stopcock
Expand
View Pricing
22145-U
Gas Sampling Bulb,
size 1000 mL, PTFE stopcock
Expand
View Pricing
22163-U
Gas Sampling Bulb,
size 500 mL, PTFE stopcock
Expand
View Pricing
22147-U
Gas Sampling Bulb,
size 250 mL, glass stopcock
Expand
View Pricing
22146-U
Gas Sampling Bulb,
size 125 mL, glass stopcock
Expand
View Pricing