
Fraxin

Synonym(s):
7,8-Dihydroxy-6-methoxycoumarin 8-β-D-glucopyranoside, 8-(β-D-Glucopyranosyloxy)-7-hydroxy-6-methoxy-2H-1-benzopyran-2-one, 8-(β-D-Glucopyranosyloxy)-7-hydroxy-6-methoxycoumarin, Fraxetin 8-β-D-glucopyranoside, Fraxetol-8-glucoside, Fraxoside
Empirical Formula (Hill Notation):
C16H18O10
CAS Number:
Molecular Weight:
370.31
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing