Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
FORTUNA
®
Optifix
®
Solvent bottle-top dispenser
Products
Product Category
dispensers (9)
Brand
Sigma-Aldrich (6)
Manufacturer
Volume (ml)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z260126
FORTUNA
®
Optifix
®
Solvent bottle-top dispenser,
Solvents model, volume 2-10 mL
Expand
View Pricing
Z260185
FORTUNA
®
Optifix
®
Solvent bottle-top dispenser,
Solvents model, volume 100-500 mL
Expand
View Pricing
Z260118
FORTUNA
®
Optifix
®
Solvent bottle-top dispenser,
Solvents model, volume 1-5 mL
Expand
View Pricing
Z260142
FORTUNA
®
Optifix
®
Solvent bottle-top dispenser,
Solvents model, volume 10-50 mL
Expand
View Pricing
Z260150
FORTUNA
®
Optifix
®
Solvent bottle-top dispenser,
Solvents model, volume 20-100 mL
Expand
View Pricing
Z260169
FORTUNA
®
Optifix
®
Solvent bottle-top dispenser,
Solvents model, volume 40-200 mL
Expand
View Pricing
Z260177
FORTUNA
®
Optifix
®
Solvent bottle-top dispenser,
Solvents model, volume 60-300 mL
Expand
View Pricing
Z260096
FORTUNA
®
Optifix
®
Solvent bottle-top dispenser,
Solvents model, volume 0.4-2.0 mL
Expand
View Pricing
Z260134
FORTUNA
®
Optifix
®
Solvent bottle-top dispenser,
Solvents model, volume 6-30 mL
Expand
View Pricing