Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fold and Snap test tube rack
Products
Product Category
racks (10)
Color
Manufacturer
Height (mm)
Length (mm)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z693022
Fold and Snap test tube rack,
for tubes, 17 mm, blue
Expand
View Pricing
Z693030
Fold and Snap test tube rack,
for tubes, 17 mm, white
Expand
View Pricing
Z692980
Fold and Snap test tube rack,
for tubes, 13 mm, blue
Expand
View Pricing
Z693065
Fold and Snap test tube rack,
for tubes, 21 mm, blue
Expand
View Pricing
Z693057
Fold and Snap test tube rack,
for tubes, 17 mm, yellow
Expand
View Pricing
Z693073
Fold and Snap test tube rack,
for tubes, 21 mm, white
Expand
View Pricing
Z693103
Fold and Snap test tube rack,
for tubes, 21 mm, yellow
Expand
View Pricing
Z693006
Fold and Snap test tube rack,
for tubes, 13 mm, red
Expand
View Pricing
Z693014
Fold and Snap test tube rack,
for tubes, 13 mm, yellow
Expand
View Pricing
Z693081
Fold and Snap test tube rack,
for tubes, 21 mm, red
Expand
View Pricing