Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Flash Point Standard - Pensky Martens
Products
Manufacturer
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (6)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
FPPMCC1
Flash Point Standard - Pensky Martens,
certified reference material, ASTM D 93 Proc. A (59.0 °C)
Expand
View Pricing
FPPMCC6
Flash Point Standard - Pensky Martens,
certified reference material, ASTM D 93 Proc. A (232.0 °C)
Expand
View Pricing
FPPMCC2
Flash Point Standard - Pensky Martens,
certified reference material, ASTM D 93 Proc. A (76.5 °C)
Expand
View Pricing
FPPMCC3
Flash Point Standard - Pensky Martens,
certified reference material, ASTM D 93 Proc. A (116.0 °C)
Expand
View Pricing
FPPMCC4
Flash Point Standard - Pensky Martens,
certified reference material, ASTM D 93 Proc. A (143.0 °C)
Expand
View Pricing
FPPMCC5
Flash Point Standard - Pensky Martens,
certified reference material, ASTM D 93 Proc. A (184.0 °C)
Expand
View Pricing