Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Flash Point Standard
EC Number:
265-149-8
Products
Manufacturer
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (7)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
CRMUPMCCMID
Flash Point Standard,
PMCC ASTM D 93 Proc A (141.0 °C)
Expand
View Pricing
CRMUPMGO
Flash Point Standard,
PMCC ASTM D 93 Proc A (57.3 °C)
Expand
View Pricing
CRMUPMCCLOW
Flash Point Standard,
PMCC ASTM D 93 Proc A (77.5 °C)
Expand
View Pricing
CRMUABKR
Flash Point Standard,
Abel IP 170 (41.2 °C)
Expand
View Pricing
CRMUPMCCHIGH
Flash Point Standard,
PMCC ASTM D 93 Proc A (228.0 °C)
Expand
View Pricing
CRMUFCLU
Flash Point Standard,
COC ASTM D 92 (256.8 °C)
Expand
View Pricing
CRMTAKR
Flash Point Standard,
TAG ASTM D56 (43.1 °C)
Expand
View Pricing
CRMUCOCLOW
Flash Point Standard,
COC ASTM D 92 (85.0 °C)
Expand
View Pricing
CRMUPMLUB
Flash Point Standard,
PMCC ASTM D 93 Proc B (105.7 °C)
Expand
View Pricing
CRMABKR
Flash Point Standard,
Abel IP 170 (41.9 °C)
Expand
View Pricing
CRMUPMLU
Flash Point Standard,
PMCC ASTM D 93 Proc B (190.5 °C)
Expand
View Pricing