Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Finnpipette
®
pipette tips
Products
Product Category
pipette tips (9)
Brand
Sigma-Aldrich (4)
Manufacturer
Length (cm)
Volume Range (µL)
Color
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
P2924
Finnpipette
®
pipette tips,
Finntip 5 mL, non-sterile
Expand
View Pricing
P7049
Finnpipette
®
pipette tips,
Finntip 250 universal, non-sterile
Expand
View Pricing
P2674
Finnpipette
®
pipette tips,
Finntip 1000, non-sterile
Expand
View Pricing
P3049
Finnpipette
®
pipette tips,
Finntip 250 Universal, non-sterile
Expand
View Pricing
P7174
Finnpipette
®
pipette tips,
Finntip 1000, non-sterile
Expand
View Pricing
P8979
Finnpipette
®
pipette tips,
Finntip 250 universal, sterile
Expand
View Pricing
P2549
Finnpipette
®
pipette tips,
Finntip 200 extended, non-sterile
Expand
View Pricing
P9560
Finnpipette
®
pipette tips,
Finntip 300, non-sterile
Expand
View Pricing
P9685
Finnpipette
®
pipette tips,
Finntip 300, non-sterile
Expand
View Pricing