
Fentanyl Related Compound E

Synonym(s):
N-Phenyl-1-(2-phenylethyl)-4-piperidinamine, N-[1-(2-Phenylethyl)-4-piperidinyl]aniline, Despropionylfentanyl
Empirical Formula (Hill Notation):
C19H24N2
CAS Number:
Molecular Weight:
280.41
Products
Brand
Formula Weight
Agency
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing