
Fentanyl citrate

Synonym(s):
Fentanyl citrate salt
Empirical Formula (Hill Notation):
C22H28N2O · C6H8O7
CAS Number:
Molecular Weight:
528.59
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing