Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Feed Water Conductivity Cell
Synonym(s):
Feed Water Conductivity Cell
Products
Product Category
sensors (3)
Brand
Milli-Q (3)
Compatibility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
ZFC0NDCA1
Feed Water Conductivity Cell,
For use with Milli-Q
®
IQ 7000, To monitor the ionic quality of feed water
Expand
View Pricing
ZEQ7C0NDC
Feed Water Conductivity Cell,
For use with Milli-Q
®
EQ 7000 water purification systems, monitors the ionic quality of feed water
Expand
View Pricing
ZFC0NDCL1
Feed Water Conductivity Cell,
for use with Milli-Q
®
Reference, To monitor the ionic quality of feed water
Expand
View Pricing