
Ethylmagnesium chloride solution

Linear Formula:
CH3CH2MgCl
CAS Number:
Molecular Weight:
88.82
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing