Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ethyl cellulose
CAS Number:
9004-57-3
Products
Product Category
carbohydrates (2)
Brand
Sigma-Aldrich (7)
Color
Manufacturer
Physical Form
Format
Application
Polymer Composition
Polymer Type
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (2)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
46080
Ethyl cellulose,
48.0-49.5% (w/w) ethoxyl basis
Expand
View Pricing
46070
Ethyl cellulose,
48.0-49.5% (w/w) ethoxyl basis
Expand
View Pricing
433837
Ethyl cellulose,
viscosity 46 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Expand
View Pricing
247499
Ethyl cellulose,
viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Expand
View Pricing
200646
Ethyl cellulose,
viscosity 4 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Expand
View Pricing
200654
Ethyl cellulose,
viscosity 300 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Expand
View Pricing
200689
Ethyl cellulose,
viscosity 10 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Expand
View Pricing
200697
Ethyl cellulose,
viscosity 22 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Expand
View Pricing
1265504
Ethyl cellulose,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
Y0001633
Ethyl cellulose,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing