
Ethyl 1-aminocyclopropanecarboxylate hydrochloride

Synonym(s):
1-Aminocyclopropyl carboxylic acid ethyl ester hydrochloride
Empirical Formula (Hill Notation):
C6H12ClNO2
CAS Number:
Molecular Weight:
165.62
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid Type
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing