Skip to Content
Merck
Ethoprophos PESTANAL®, analytical standard

Ethoprophos

Ethoprophos PESTANAL®, analytical standard
Synonym(s):
O-Ethyl S,S-dipropyl phosphorodithioate, Ethoprofos, Ethoprop
Empirical Formula (Hill Notation):
C8H19O2PS2
CAS Number:
Molecular Weight:
242.34
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing