
Estradiol benzoate

Synonym(s):
β-Estradiol 3-benzoate, 1,3,5(10)-Estratriene-3,17β-diol 3-benzoate, 3,17β-Dihydroxy-1,3,5(10)-estratriene 3-benzoate, 3-Benzoyloxy-17β-estrol
Empirical Formula (Hill Notation):
C25H28O3
CAS Number:
Molecular Weight:
376.49
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing