Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Xplorer
®
Plus single channel pipettor
Products
Product Category
pipettors (5)
Color
Manufacturer
Volume (ml)
Volume (µL)
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
EP4861000716
Eppendorf
®
Xplorer
®
Plus single channel pipettor,
multifunction rocker switch, yellow, volume 5-100 μL, for use with for pipette tips 200 mL
Expand
View Pricing
EP4861000732
Eppendorf
®
Xplorer
®
Plus single channel pipettor,
volume 50-1000 μL, multifunction rocker switch, blue, for use with fpr pipette tips 1000 mL
Expand
View Pricing
EP4861000740
Eppendorf
®
Xplorer
®
Plus single channel pipettor,
volume 0.2-5 mL, multifunction rocker switch, purple, for pipette tips 5 mL
Expand
View Pricing
EP4861000759
Eppendorf
®
Xplorer
®
Plus single channel pipettor,
volume 0.5-10 mL, multifunction rocker switch, turquoise, for pipette tips 10 mL
Expand
View Pricing
EP4861000708
Eppendorf
®
Xplorer
®
Plus single channel pipettor,
volume (0.5-10 μL), multifunction rocker switch, medium gray, for pipette tips 20 mL
Expand
View Pricing