Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Twin tec microbiology PCR plates, 96 wells
Products
Color
Manufacturer
Wells
Color
Package Size
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
EP0030129326
Eppendorf
®
Twin tec microbiology PCR plates, 96 wells,
colorless plate, 96, semi-skirted, conical bottom colorless wells, PCR clean, semi-skirted, pkg of 10 ea, colorless
Expand
View Pricing
EP0030129334
Eppendorf
®
Twin tec microbiology PCR plates, 96 wells,
blue plate, 96, semi-skirted, conical bottom colorless wells, semi-skirted, PCR clean, blue, pkg of 10 ea
Expand
View Pricing