Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Research
®
Plus Move It pipette
Synonym(s):
Eppendorf
®
Research
®
Plus Move It pipette
Products
Product Category
pipettors (10)
Manufacturer
Volume (µL)
Color
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
EP3125000206
Eppendorf
®
Research
®
Plus Move It pipette,
no. of channels, 8, volume 120-1200 μL, cone distance 4.5-14 mm, dark green
Expand
View Pricing
EP3125000125
Eppendorf
®
Research
®
Plus Move It pipette,
no. of channels, 12, volume 0.1-20 μL, 4.5-9 mm cone distance, light pink
Expand
View Pricing
EP3125000176
Eppendorf
®
Research
®
Plus Move It pipette,
no. of channels, 8, volume 30-300 μL, cone distance 4.5-14 mm, orange
Expand
View Pricing
EP3125000192
Eppendorf
®
Research
®
Plus Move It pipette,
no. of channels, 6, volume 120-1200 μL, cone distance 9-20 mm, dark green
Expand
View Pricing
EP3125000150
Eppendorf
®
Research
®
Plus Move It pipette,
no. of channels, 4, volume 30-300 μL, cone distance 9-33 mm, orange
Expand
View Pricing
EP3125000141
Eppendorf
®
Research
®
Plus Move It pipette,
no. of channels, 12, volume 0.5-100 μL, 4.5-9 mm cone distance, light yellow
Expand
View Pricing
EP3125000117
Eppendorf
®
Research
®
Plus Move It pipette,
no. of channels, 8, volume 0.1-20 μL, 4.5-14 mm cone distance, light pink
Expand
View Pricing
EP3125000133
Eppendorf
®
Research
®
Plus Move It pipette,
no. of channels, 8, volume 0.5-100 μL, 4.5-14 mm cone distance, light yellow
Expand
View Pricing
EP3125000184
Eppendorf
®
Research
®
Plus Move It pipette,
no. of channels, 4, volume 120-1200 μL, cone distance 9-33 mm, dark green
Expand
View Pricing
EP3125000168
Eppendorf
®
Research
®
Plus Move It pipette,
no. of channels, 6, volume 30-300 μL, cone distance 9-20 mm, orange
Expand
View Pricing