Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Reference 2 G, BASIC, single-channel, variable
Products
Product Category
pipettors (1)
Color
Manufacturer
Volume (ml)
Volume (µL)
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
EP4924000223
Eppendorf
®
Reference 2 G, BASIC, single-channel, variable,
volume 0.5-10 μL, medium gray
Expand
View Pricing
EP4924000215
Eppendorf
®
Reference 2 G, BASIC, single-channel, variable,
volume 0.1-2.5 μL, dark gray
Expand
View Pricing
EP4924000282
Eppendorf
®
Reference 2 G, BASIC, single-channel, variable,
volume 100-1000 μL, blue
Expand
View Pricing
EP4924000266
Eppendorf
®
Reference 2 G, BASIC, single-channel, variable,
volume 20-200 μL, yellow
Expand
View Pricing
EP4924000070
Eppendorf
®
Reference 2 G, BASIC, single-channel, variable,
volume 30-300 μL, orange, incl. epTIPS box
Expand
View Pricing
EP4924000231
Eppendorf
®
Reference 2 G, BASIC, single-channel, variable,
volume 2-20 μL, light gray
Expand
View Pricing
EP4924000304
Eppendorf
®
Reference 2 G, BASIC, single-channel, variable,
volume 0.5-5 mL, purple
Expand
View Pricing