Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Multipette
®
M4
Synonym(s):
Eppendorf
®
Multipette
®
M4
Products
Product Category
pipettors (3)
Brand
Sigma-Aldrich (2)
Manufacturer
Volume (ml)
Volume (µL)
Color
Package Size
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
EP4982000012
Eppendorf
®
Multipette
®
M4,
with holder
Expand
View Pricing
EP4982000411
Eppendorf
®
Multipette
®
M4,
Includes holder for wall mount or pipette carousel
Expand
View Pricing
EP4982000314
Eppendorf
®
Multipette
®
M4,
starter kit, adjustable
Expand
View Pricing