Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Fixed-angle rotor
Products
Product Category
rotors (11)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
EP5804727002
Eppendorf
®
Fixed-angle rotor,
F-34-6-38, 6 x 85 ml incl. lid
Expand
View Pricing
EP5804726006
Eppendorf
®
Fixed-angle rotor,
FA-45-30-11, 30x 1,5/2 ml, incl. aluminium lid, aerosol-tight
Expand
View Pricing
EP5427753001
Eppendorf
®
Fixed-angle rotor,
FA-45-30-11, incl. rotor lid, aerosol-tight
Expand
View Pricing
EP5804715004
Eppendorf
®
Fixed-angle rotor,
F45-30-11 30 x 1.5/2 ml, incl. aluminium lid
Expand
View Pricing
EP5427739009
Eppendorf
®
Fixed-angle rotor,
F-35-6-30, incl. rotor lid
Expand
View Pricing
EP5702746002
Eppendorf
®
Fixed-angle rotor,
F-45-24-11, without lid
Expand
View Pricing
EP5702704008
Eppendorf
®
Fixed-angle rotor,
F-35-30-17, 30 x 15 ml, incl. 20 adapters Falcon tubes and 30 rubber mats
Expand
View Pricing
EP5427752005
Eppendorf
®
Fixed-angle rotor,
FA-45-24-11 -Kit, incl. rotor lid, aerosol -tight
Expand
View Pricing
EP5804735005
Eppendorf
®
Fixed-angle rotor,
to hold, 6 x 8-tube or 5-tube PCR strips, to hold, 48 x 0.2 mL PCR tubes
Expand
View Pricing
EP5702705004
Eppendorf
®
Fixed-angle rotor,
F-35-30-17, 10 x 15 ml, incl. 10 adapters Falcon tubes and 10 rubber mats
Expand
View Pricing
EP5702747009
Eppendorf
®
Fixed-angle rotor,
F-45-18-17 Cryo, without lid
Expand
View Pricing