Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
epT.I.P.S.
®
G Reloads, Eppendorf Quality
™
Products
Product Category
pipette tips (9)
Color
Manufacturer
Volume (ml)
Volume (µL)
Color
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
EP0030073436
Eppendorf
®
epT.I.P.S.
®
G Reloads, Eppendorf Quality
™
,
volume (2-200 μL), L (53 mm), yellow, pack of 960 ea (10 trays × 96 tips stacked)
Expand
View Pricing
EP0030073398
Eppendorf
®
epT.I.P.S.
®
G Reloads, Eppendorf Quality
™
,
volume (0.1-20 μL), L (40 mm), medium gray, pack of 960 ea (10 trays × 96 tips)
Expand
View Pricing
EP0030073479
Eppendorf
®
epT.I.P.S.
®
G Reloads, Eppendorf Quality
™
,
volume (50-1000 μL), L (71 mm), blue, pack of 960 ea (10 trays × 96 tips)
Expand
View Pricing
EP0030073371
Eppendorf
®
epT.I.P.S.
®
G Reloads, Eppendorf Quality
™
,
volume (0.1-10 μL), L (34 mm), dark gray, pack of 960 ea (10 trays × 96 tips, stacked)
Expand
View Pricing
EP0030073452
Eppendorf
®
epT.I.P.S.
®
G Reloads, Eppendorf Quality
™
,
volume (20-300 μL), L (55 mm), orange, pack of 960 ea (10 trays × 96 tips)
Expand
View Pricing
EP0030073495
Eppendorf
®
epT.I.P.S.
®
G Reloads, Eppendorf Quality
™
,
volume (50-1250 μL), L (76 mm), green, pack of 960 ea (10 trays × 96 tips)
Expand
View Pricing
EP0030073410
Eppendorf
®
epT.I.P.S.
®
G Reloads, Eppendorf Quality
™
,
volume (0.5-20 μL), L (46 mm), light gray, pack of 960 ea (10 trays × 96 tips)
Expand
View Pricing
EP0030073630
Eppendorf
®
epT.I.P.S.
®
G Reloads, Eppendorf Quality
™
,
volume (50-1250 μL), L (103 mm), dark green, pack of 960 ea (10 trays × 96 tips)
Expand
View Pricing
EP0030073517
Eppendorf
®
epT.I.P.S.
®
G Reloads, Eppendorf Quality
™
,
volume (0.25-2.5 mL), L (115 mm), red, pack of 480 ea (10 trays × 48 tips)
Expand
View Pricing