Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
epT.I.P.S.
®
Biopur
Synonym(s):
Eppendorf
®
epT.I.P.S.
®
Biopur
Products
Product Category
pipette tips (10)
Color
Manufacturer
Volume Range (µL)
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
EP0030075234
Eppendorf
®
epT.I.P.S.
®
Biopur,
tip volume range × L 2-200 μL × 53 mm, yellow tip, case of 480 ea (5 racks x 96 tips)
Expand
View Pricing
EP0030075315
Eppendorf
®
epT.I.P.S.
®
Biopur,
tip volume range × L 0.5-10 mL × 165 mm, turquoise tip, case of 120 ea (5 racks x 24 tips)
Expand
View Pricing
EP0030075293
Eppendorf
®
epT.I.P.S.
®
Biopur,
tip volume range × L 0.1-5 mL × 120 mm, violet tip, case of 120 ea (5 racks x 24 tips)
Expand
View Pricing
EP0030075250
Eppendorf
®
epT.I.P.S.
®
Biopur,
tip volume range × L 50-1000 μL × 71 mm, blue tip, case of 480 ea (5 racks x 96 tips)
Expand
View Pricing
EP0030075226
Eppendorf
®
epT.I.P.S.
®
Biopur,
tip volume range × L 0.1-20 μL × 40 mm, medium gray tip, case of 480 ea (5 racks x 96 tips)
Expand
View Pricing
EP0030075242
Eppendorf
®
epT.I.P.S.
®
Biopur,
tip volume range × L 2-300 μL × 55 mm, orange tip, case of 480 ea (5 racks x 96 tips)
Expand
View Pricing
EP0030075285
Eppendorf
®
epT.I.P.S.
®
Biopur,
tip volume range × L 0.25-2.5 mL × 115 mm, red tip, case of 240 ea (5 racks x 48 tips)
Expand
View Pricing
EP0030075307
Eppendorf
®
epT.I.P.S.
®
Biopur,
tip volume range × L 0.2-5 mL × 175 mm, violet, pack of 120 ea (5 racks x 24 tips)
Expand
View Pricing
EP0030075269
Eppendorf
®
epT.I.P.S.
®
Biopur,
tip volume range × L 50-1250 μL × 76 mm, green tip, case of 480 ea (5 racks x 96 tips)
Expand
View Pricing
EP0030075277
Eppendorf
®
epT.I.P.S.
®
Biopur,
tip volume range × L 50-1250 μL × 103 mm, dark green tip, case of 480 ea (5 racks x 96 tips)
Expand
View Pricing