Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Conical Tubes
Products
Product Category
tubes (9)
vials (1)
Brand
Sigma-Aldrich (1)
Color
Feature
Manufacturer
Capacity (mL)
Diameter (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
EP0030122178
Eppendorf
®
Conical Tubes,
sterile, 50 mL, colorless, pkg of 500 tubes (10 bags of 50 tubes)
Expand
View Pricing
EP0030122151
Eppendorf
®
Conical Tubes,
sterile, 15 mL, colorless, pkg of 500 tubes (10 bags of 50 tubes)
Expand
View Pricing
EP0030122186
Eppendorf
®
Conical Tubes,
sterile, pyrogen-, DNase-, RNase-, and DNA-free, pack of 300 ea (12 racks x 25 tubes)
Expand
View Pricing
EP0030122160
Eppendorf
®
Conical Tubes,
sterile, pyrogen-, DNase-, RNase-, and DNA-free, pack of 500 ea (20 racks x 25 tubes)
Expand
View Pricing
EP0030122259
Eppendorf
®
Conical Tubes,
Forensic DNA grade, 15 mL, colorless, pkg of 100 ea (individually wrapped)
Expand
View Pricing
EP0030122267
Eppendorf
®
Conical Tubes,
Forensic DNA grade, 50 mL, colorless, pkg of 100 ea (individually wrapped)
Expand
View Pricing
EP0030122445
Eppendorf
®
Conical Tubes,
sterile, screw top cap, endotoxin free, Biopur, DNase free, Biopur, capacity 25 mL, amber, pkg of 200 ea (8 bags x 25 tubes)
Expand
View Pricing
EP0030118430
Eppendorf
®
Conical Tubes,
sterile, cap (snapTech ), endotoxin free, Biopur, DNase free, Biopur, capacity 25 mL, amber, pkg of 150 ea (6 bags x 25 tubes)
Expand
View Pricing
EP0030113519
Eppendorf
®
Conical Tubes,
Forensic DNA Grade, volume 50 mL, colorless (with 2D SafeCode), pkg of 48 tubes (individually wrapped)
Expand
View Pricing