Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Combitips Advanced
®
Products
Product Category
pipette tips (33)
Color
Manufacturer
Volume (ml)
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-20 of 33
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
EP0030089464
Eppendorf
®
Combitips Advanced
®
,
volume 10 mL, non-sterile, orange, pack of 100 ea
Expand
View Pricing
EP0030089456
Eppendorf
®
Combitips Advanced
®
,
volume (5 mL), blue, pack of 100 ea
Expand
View Pricing
EP0030089421
Eppendorf
®
Combitips Advanced
®
,
volume (0.5 mL), purple, pack of 100 ea
Expand
View Pricing
EP0030089430
Eppendorf
®
Combitips Advanced
®
,
volume (1.0 mL), yellow
Expand
View Pricing
EP0030089448
Eppendorf
®
Combitips Advanced
®
,
volume (2.5 mL), green, pack of 100 ea
Expand
View Pricing
EP0030089669
Eppendorf
®
Combitips Advanced
®
,
volume (5 mL), blue, Biopur
®
, pack of 100 ea (individually wrapped)
Expand
View Pricing
EP0030089405
Eppendorf
®
Combitips Advanced
®
,
volume (0.1 mL), white, pack of 100 ea
Expand
View Pricing
EP0030089677
Eppendorf
®
Combitips Advanced
®
,
volume (10 mL), orange, Biopur
®
, pack of 100 ea (individually wrapped)
Expand
View Pricing
EP0030089472
Eppendorf
®
Combitips Advanced
®
,
volume (25 mL), red, pack of 100 ea
Expand
View Pricing
EP0030089413
Eppendorf
®
Combitips Advanced
®
,
volume (0.2 mL), light blue, pack of 100 ea
Expand
View Pricing
EP0030089480
Eppendorf
®
Combitips Advanced
®
,
volume (50 mL), light gray, pack of 100 ea
Expand
View Pricing
EP0030089650
Eppendorf
®
Combitips Advanced
®
,
volume (2.5 mL), green, Biopur
®
, pack of 100 ea (individually wrapped)
Expand
View Pricing
EP0030089642
Eppendorf
®
Combitips Advanced
®
,
volume (1.0 mL), yellow, Biopur
®
, pack of 100 ea (individually wrapped)
Expand
View Pricing
EP0030089685
Eppendorf
®
Combitips Advanced
®
,
volume (25 mL), red, Biopur
®
, pack of 100 ea (individually wrapped)
Expand
View Pricing
EP0030089634
Eppendorf
®
Combitips Advanced
®
,
volume (0.5 mL), purple, Biopur
®
, pack of 100 ea (individually wrapped)
Expand
View Pricing
EP0030089693
Eppendorf
®
Combitips Advanced
®
,
volume (50 mL), light gray, Biopur
®
, pack of 100 ea (individually wrapped)
Expand
View Pricing
EP0030089618
Eppendorf
®
Combitips Advanced
®
,
volume (0.1 mL), white, Biopur
®
, pack of 100 ea (individually wrapped)
Expand
View Pricing
EP0030089782
Eppendorf
®
Combitips Advanced
®
,
volume (0.5 mL), purple, PCR clean, pack of 100 ea (4 reclosable bags of 25 tips)
Expand
View Pricing
EP0030089774
Eppendorf
®
Combitips Advanced
®
,
volume (0.2 mL), light blue, PCR clean, pack of 100 ea (4 reclosable bags of 25 tips)
Expand
View Pricing
EP0030089626
Eppendorf
®
Combitips Advanced
®
,
volume (0.2 mL), light blue, Biopur
®
, pack of 100 ea (individually wrapped)
Expand
View Pricing
Page 1 of 2
Page 1 of 2