Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Centrifuge 5425 R
Products
Product Category
centrifuges (8)
Feature
Manufacturer
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
EP5406000518
Eppendorf
®
Centrifuge 5425 R,
rotory knob control, with rotor FA-24x2, AC/DC input 230 V (EU Plug)
Expand
View Pricing
EP5406000119
Eppendorf
®
Centrifuge 5425 R,
keypad, with rotor FA-24x2, refrigerated, AC/DC input 230 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP5406000160
Eppendorf
®
Centrifuge 5425 R,
keypad control, with rotor FA-24x2, AC/DC input 230 V (GB Plug)
Expand
View Pricing
EP5406000569
Eppendorf
®
Centrifuge 5425 R,
rotory knob control, with rotor FA-24x2, AC/DC input 230 V (GB Plug)
Expand
View Pricing
EP5406000313
Eppendorf
®
Centrifuge 5425 R,
keypad control, without rotor, AC/DC input 230 V (EU Plug)
Expand
View Pricing
EP5406000364
Eppendorf
®
Centrifuge 5425 R,
keypad control, without rotor, AC/DC input 230 V (GB Plug)
Expand
View Pricing
EP5406000712
Eppendorf
®
Centrifuge 5425 R,
rotory knob control, without rotor, AC/DC input 230 V (EU Plug)
Expand
View Pricing
EP5406000763
Eppendorf
®
Centrifuge 5425 R,
rotory knob control, without rotor, AC/DC input 230 V (GB Plug)
Expand
View Pricing